Learn More
Medchemexpress LLC Aclacinomycin A | 57576-44-0 | 1 ML
SDP
Catalog No. 5000421362
Encompass_Preferred
Aclacinomycin A (Aclarubicin) is an orally active and potent anthracycline antitumor antibiotic. It acts as an inhibitor of topoisomerase I and II, inhibiting the synthesis of nucleic acid, particularly RNA. It may also inhibit the 26S protease complex and ubiquitin-ATP-dependent proteolysis.
  • Orally active and potent anthracycline antitumor antibiotic.
  • Inhibitor of topoisomerase I and II.
  • Inhibits nucleic acid synthesis, especially RNA.
  • May inhibit 26S protease complex and ubiquitin-ATP-dependent proteolysis.
  • Exhibits antitumor effect on leukemia L-1210.
  • Well absorbed orally in mice, rats, and dogs.
Catalog No. 50-004-21362 Supplier Medchemexpress LLC Supplier No. HYN230610MM/1ML
Aclacinomycin A (Aclarubicin) is an orally active and potent anthracycline antitumor antibiotic. It acts as an inhibitor of topoisomerase I and II, inhibiting the synthesis of nucleic acid, particularly RNA. It may also inhibit the 26S protease complex and ubiquitin-ATP-dependent proteolysis.
  • Orally active and potent anthracycline antitumor antibiotic.
  • Inhibitor of topoisomerase I and II.
  • Inhibits nucleic acid synthesis, especially RNA.
  • May inhibit 26S protease complex and ubiquitin-ATP-dependent proteolysis.
  • Exhibits antitumor effect on leukemia L-1210.
  • Well absorbed orally in mice, rats, and dogs.