Learn More
Enzo Life Sciences Aclacinomycin A, (5mg), CAS Number: 57576-44-0
SDP
Catalog No. NC1679530
Encompass_Preferred
Originally identified as an anti-tumor drug, aclacinomycin A is the first described non-peptidic inhibitor showing discrete specificity for the chymotrypsin-like activity of the 20S proteasome. Aclacinomycin A has no effect on cathepsin B, it stimulates trypsin activity but inhibits the activity of both chymotrypsin and calpain. Alternative Name: Aclarubicin. Formula: C42H53NO15. MW: 811.9. CAS Number: 57576-44-0. Purity: ≥95% (TLC). Appearance: Yellow powder with orange cast. Solubility: Soluble in DMSO (25mg/ml) or dimethyl formamide (25mg/ml). Long Term Storage: -20°C
Catalog No. NC1679530 Supplier Enzo Life Sciences Supplier No. BMLAW86550005
Originally identified as an anti-tumor drug, aclacinomycin A is the first described non-peptidic inhibitor showing discrete specificity for the chymotrypsin-like activity of the 20S proteasome. Aclacinomycin A has no effect on cathepsin B, it stimulates trypsin activity but inhibits the activity of both chymotrypsin and calpain. Alternative Name: Aclarubicin. Formula: C42H53NO15. MW: 811.9. CAS Number: 57576-44-0. Purity: ≥95% (TLC). Appearance: Yellow powder with orange cast. Solubility: Soluble in DMSO (25mg/ml) or dimethyl formamide (25mg/ml). Long Term Storage: -20°C