Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aclacinomycin A (Aclarubicin) | 57576-44-0 | 1 MG
SDP

Catalog No. 5000420405
Encompass_Preferred

Aclacinomycin A (Aclarubicin) is an orally active and potent anthracycline antitumor antibiotic. It functions as an inhibitor of topoisomerase I and II, and it also inhibits the synthesis of nucleic acids, primarily RNA. Furthermore, it may inhibit the 26S protease complex and ubiquitin-ATP-dependent proteolysis.

  • Acts as an inhibitor of topoisomerase I and II
  • Inhibits the synthesis of nucleic acids, particularly RNA
  • May inhibit the 26S protease complex
  • May inhibit ubiquitin-ATP-dependent proteolysis
  • Orally active
  • Potent anthracycline antitumor antibiotic
  • Assay purity: 99.30%
  • Molecular weight: 811.87
  • Chemical formula: C42H53NO15
  • Appearance: Light yellow to yellow solid
  • Isolated from microorganisms, Streptomyces sp.

Catalog No. 50-004-20405 Supplier Medchemexpress LLC Supplier No. HYN23061MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aclacinomycin A (Aclarubicin) is an orally active and potent anthracycline antitumor antibiotic. It functions as an inhibitor of topoisomerase I and II, and it also inhibits the synthesis of nucleic acids, primarily RNA. Furthermore, it may inhibit the 26S protease complex and ubiquitin-ATP-dependent proteolysis.

  • Acts as an inhibitor of topoisomerase I and II
  • Inhibits the synthesis of nucleic acids, particularly RNA
  • May inhibit the 26S protease complex
  • May inhibit ubiquitin-ATP-dependent proteolysis
  • Orally active
  • Potent anthracycline antitumor antibiotic
  • Assay purity: 99.30%
  • Molecular weight: 811.87
  • Chemical formula: C42H53NO15
  • Appearance: Light yellow to yellow solid
  • Isolated from microorganisms, Streptomyces sp.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.