Learn More
Medchemexpress LLC Aclidinium Bromide | 320345-99-1 | 98.96% | C26H30BrNO4S2 | 1 ML
SDP
Catalog No. 5000356777
Encompass_Preferred
Aclidinium Bromide is a long-acting, inhaled muscarinic antagonist with potential for research into chronic obstructive pulmonary disease (COPD). It has shown potent antagonism of human muscarinic receptors preclinically, with a long residence time at M3 receptors, suggesting potential for sustained bronchodilation. Early clinical studies have indicated low systemic bioavailability and a favorable safety profile.
  • Long-acting muscarinic antagonist
  • Potential for chronic obstructive pulmonary disease (COPD) research
  • Exhibits potent antagonism of human muscarinic receptors
  • Long residence time at M3 receptors for sustained bronchodilation
  • Low systemic bioavailability
  • Favorable safety profile
  • Rapidly hydrolyzed in human plasma
Catalog No. 50-003-56777 Supplier Medchemexpress LLC Supplier No. HY1414410MM/1ML
Aclidinium Bromide is a long-acting, inhaled muscarinic antagonist with potential for research into chronic obstructive pulmonary disease (COPD). It has shown potent antagonism of human muscarinic receptors preclinically, with a long residence time at M3 receptors, suggesting potential for sustained bronchodilation. Early clinical studies have indicated low systemic bioavailability and a favorable safety profile.
  • Long-acting muscarinic antagonist
  • Potential for chronic obstructive pulmonary disease (COPD) research
  • Exhibits potent antagonism of human muscarinic receptors
  • Long residence time at M3 receptors for sustained bronchodilation
  • Low systemic bioavailability
  • Favorable safety profile
  • Rapidly hydrolyzed in human plasma