Learn More
Medchemexpress LLC Aclidinium bromide | 320345-99-1 | MFCD22683690 | 99.0% | C26H30BrNO4S2 | 10MG
SDP
Catalog No. 5000261430
Encompass_Preferred
Aclidinium bromide is a long-acting inhaled muscarinic antagonist used in respiratory research, primarily for studies of chronic obstructive pulmonary disease (COPD) and muscarinic receptor function. Supplied as the bromide salt with characterized identity and high purity, it is intended for in vitro and preclinical pharmacology applications.
  • Long-acting muscarinic antagonist for COPD and airway research.
  • High purity suitable for analytical and pharmacological studies.
  • Available as solid and DMSO solution formats for flexible use.
  • Well-characterized chemical identity and molecular data.
  • Useful as a pharmacological tool to probe muscarinic receptor-mediated bronchodilation.
Catalog No. 50-002-61430 Supplier Medchemexpress LLC Supplier No. HY1414410MG
Aclidinium bromide is a long-acting inhaled muscarinic antagonist used in respiratory research, primarily for studies of chronic obstructive pulmonary disease (COPD) and muscarinic receptor function. Supplied as the bromide salt with characterized identity and high purity, it is intended for in vitro and preclinical pharmacology applications.
  • Long-acting muscarinic antagonist for COPD and airway research.
  • High purity suitable for analytical and pharmacological studies.
  • Available as solid and DMSO solution formats for flexible use.
  • Well-characterized chemical identity and molecular data.
  • Useful as a pharmacological tool to probe muscarinic receptor-mediated bronchodilation.