Learn More
Selleck Chemical LLC Aclidinium Bromide S4031-10mg
SDP
Catalog No. 503108875
Encompass_Preferred
Aclidinium Bromide (LAS 34273 LAS-W 330) inhibits human muscarinic AChR M1 M2 M3 M4 and M5 with Ki of 0 1 nM 0 14 nM 0 14 nM 0 21 nM and 0 16 nM respectively
Catalog No. 50-310-8875 Supplier Selleck Chemical LLC Supplier No. S403110MG
Aclidinium Bromide (LAS 34273 LAS-W 330) inhibits human muscarinic AChR M1 M2 M3 M4 and M5 with Ki of 0 1 nM 0 14 nM 0 14 nM 0 21 nM and 0 16 nM respectively