Learn More
Medchemexpress LLC HY-14144 5mg Medchemexpress, Aclidinium (Bromide) CAS:320345-99-1 Purity:>98%
SDP
Catalog No. 502081565
Encompass_Preferred
Medchemexpress, HY-14144 5mg Aclidinium (Bromide) CAS:320345-99-1 Aclidinium Bromide (LAS 34273; LAS-W 330) is a long-acting, inhaled muscarinic antagonist. Aclidinium Bromide has the potential for chronic obstructive pulmonary disease (COPD) research[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1565 Supplier Medchemexpress LLC Supplier No. HY141445MG
Medchemexpress, HY-14144 5mg Aclidinium (Bromide) CAS:320345-99-1 Aclidinium Bromide (LAS 34273; LAS-W 330) is a long-acting, inhaled muscarinic antagonist. Aclidinium Bromide has the potential for chronic obstructive pulmonary disease (COPD) research[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.