Learn More
Medchemexpress LLC ACLY Human His-SUM 100ug
SDP
Catalog No. 5000195476
Encompass_Preferred
ACLY Protein, Human (His-SUMO) is a recombinant human ATP-citrate lyase produced with an N-terminal His-SUMO fusion tag and expressed in E. coli. It catalyzes the generation of acetyl-CoA from citrate and is supplied as a lyophilized, purified protein for biochemical and enzymology studies. Manufacturer certificates indicate >90% purity and an approximate molecular weight of 45.5 kDa. Store frozen and aliquot after reconstitution for best stability.
  • Recombinant human sequence expressed in E. coli.
  • N-terminal His-SUMO tag for improved solubility and purification.
  • Supplied as lyophilized powder for long-term storage.
  • High purity suitable for biochemical and enzymatic assays.
  • Stable when aliquoted and stored at -20 °C or -80 °C after reconstitution.
Catalog No. 50-001-95476 Supplier Medchemexpress LLC Supplier No. HYP72070100UG
ACLY Protein, Human (His-SUMO) is a recombinant human ATP-citrate lyase produced with an N-terminal His-SUMO fusion tag and expressed in E. coli. It catalyzes the generation of acetyl-CoA from citrate and is supplied as a lyophilized, purified protein for biochemical and enzymology studies. Manufacturer certificates indicate >90% purity and an approximate molecular weight of 45.5 kDa. Store frozen and aliquot after reconstitution for best stability.
  • Recombinant human sequence expressed in E. coli.
  • N-terminal His-SUMO tag for improved solubility and purification.
  • Supplied as lyophilized powder for long-term storage.
  • High purity suitable for biochemical and enzymatic assays.
  • Stable when aliquoted and stored at -20 °C or -80 °C after reconstitution.