Learn More
Medchemexpress LLC ACLY Human His-SUM 5ug
SDP
Catalog No. 5000196272
Encompass_Preferred
ACLY Protein, Human (His-SUMO) is a recombinant fragment of human ATP-citrate lyase (amino acids K4-K265) produced in E. coli with an N-terminal His-SUMO tag. The ~45.5 kDa protein is supplied as a purified reagent for biochemical characterization, enzyme assays, and antibody generation; batch-specific certificates of analysis are available.
  • N-terminal His-SUMO tag for affinity purification.
  • Expressed in E. coli.
  • Derived from human sequence (K4-K265).
  • Approximately 45.5 kDa molecular weight.
  • Available in small research sizes, including 5 μg.
  • Certificate of analysis available for batch verification.
  • Suitable for biochemical assays and antibody production.
Catalog No. 50-001-96272 Supplier Medchemexpress LLC Supplier No. HYP720705UG
ACLY Protein, Human (His-SUMO) is a recombinant fragment of human ATP-citrate lyase (amino acids K4-K265) produced in E. coli with an N-terminal His-SUMO tag. The ~45.5 kDa protein is supplied as a purified reagent for biochemical characterization, enzyme assays, and antibody generation; batch-specific certificates of analysis are available.
  • N-terminal His-SUMO tag for affinity purification.
  • Expressed in E. coli.
  • Derived from human sequence (K4-K265).
  • Approximately 45.5 kDa molecular weight.
  • Available in small research sizes, including 5 μg.
  • Certificate of analysis available for batch verification.
  • Suitable for biochemical assays and antibody production.