Learn More
Electron Microscopy Sciences EMS Calibration Micrometer Model ACM-1
SDP
Catalog No. 5034063
Encompass_Preferred
This ideal standard provides accurate calibration of any eyepiece, reticules, filar micrometers, or electronic measuring instrument. Features a series of ʽH' shaped fiducial images in a range of sizes from 0.1 mm through 20 mm, useful for calibration range from 1X to 1000X power.
Catalog No. 50-340-63 Supplier Electron Microscopy Sciences Supplier No. 602181PG
This ideal standard provides accurate calibration of any eyepiece, reticules, filar micrometers, or electronic measuring instrument. Features a series of ʽH' shaped fiducial images in a range of sizes from 0.1 mm through 20 mm, useful for calibration range from 1X to 1000X power.