Learn More
Sino Biological AcmNPV Envelope glycoprotein gp64 / AcmNPV-gp64 Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 40496AT3020
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant AcmNPV Envelope glycoprotein gp64 / AcmNPV-gp64 ( Catalog#40496-V08H; NP_054158.1; Met1-Thr481). AcMNPV GP64 specific IgG was purified by AcmNPV Envelope glycoprotein gp64 / AcmNPV-gp64 affinity chromatography.
Catalog No. 50-255-3710