Learn More
Sino Biological AcmNPV Envelope glycoprotein gp64 / AcmNPV-gp64 Antibody(HRP), Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 40496T62H100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant AcmNPV Envelope glycoprotein gp64 / AcmNPV-gp64 ( Catalog#40496-V08B; NP_054158.1; Met1-Thr481). AcmNPV Envelope glycoprotein gp64 / AcmNPV-gp64 specific IgG was purified by AcmNPV Envelope glycoprotein gp64 / AcmNPV-gp64 affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-224-6467