Learn More
Sino Biological AcmNPV Envelope glycoprotein gp64 ELISA Pair Set
Supplier:  Sino Biological SEK4049615
Encompass_Preferred
The AcmNPV Envelope glycoprotein gp64 ELISA Pair Set is for the quantitative determination of AcmNPV Envelope glycoprotein gp64.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for AcmNPV Envelope glycoprotein gp64 coated on a 96-well plate. Standards and samples are added to the wells, and any AcmNPV Envelope glycoprotein gp64 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-AcmNPV Envelope glycoprotein gp64 monoclonal antibody is then added, producing an antibody-antigen-antibody ""sandwich"". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of AcmNPV Envelope glycoprotein gp64 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-237-2021