Learn More
Sino Biological AcMNPV-GP64 ELISA Pair Set 5PP (Sino Biological)
Catalog No. 502372022
Encompass_Preferred
Each vial contains 200 ng/mL of recombinant AcmNPV Envelope glycoprotein gp64. Store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 5000 pg/mL is recommended.
Catalog No. 50-237-2022 Supplier Sino Biological Supplier No. SEK404965
Each vial contains 200 ng/mL of recombinant AcmNPV Envelope glycoprotein gp64. Store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 5000 pg/mL is recommended.