Learn More
Cayman Chemical AcMNPV Major Envelope Glycopr
SDP
Supplier:  Cayman Chemical 37014500 UG
Encompass_Preferred
Immunogen: Recombinant AcMNPV (strain E2) • Host: Mouse • Species Reactivity: (+) AcMNPV • Cross Reactivity: (+) Major envelope glycoprotein • Applications: MN
Catalog No. 50-291-5347