Learn More
Cayman Chemical AcMNPV Major Envelope Glycopr
SDP
Supplier:  Cayman Chemical 37024100 UG
Encompass_Preferred
Source: Recombinant AcMNPV (strain E2) C-terminal His-tagged major envelope glycoprotein expressed in insect cells • Amino acids: 21-481 • MW: 54.2 kDa
Catalog No. 50-291-4828