Learn More
Cayman Chemical Ac-Nle-Pro-Nle-Asp-AMC (trifluoroacetate salt), Purity: ≥98%, 5 mg
SDP
Catalog No. 502987582
Encompass_Preferred
A substrate for proteasomes (specific activities of 113 and 6.6 nmol/min/mg by the 26S (rabbit muscle) and 20S (yeast) proteasomes, respectively); cleaved by PGPH; allosterically inhibits the chymotrypsin-like activity of the proteasome
Catalog No. 50-298-7582 Supplier Cayman Chemical Supplier No. 216395 MG
A substrate for proteasomes (specific activities of 113 and 6.6 nmol/min/mg by the 26S (rabbit muscle) and 20S (yeast) proteasomes, respectively); cleaved by PGPH; allosterically inhibits the chymotrypsin-like activity of the proteasome