Learn More
Cayman Chemical AcnthosIde B 10mg
SDP
Catalog No. 502942838
Encompass_Preferred
A lignan with diverse biological activities; scavenges DPPH and ABTS radicals in cell-free assays (IC50s = 9.9 and 12.7 µM, respectively); inhibits LPS-induced NO production in RAW 264.7 cells at 20 µM; reverses scopolamine-induced memory impairment, reduces hippocampal MDA levels, and increases hippocampal SOD, catalase, and GPX levels in a mouse model of scopolamine-induced amnesia at 20 mg/kg
Catalog No. 50-294-2838 Supplier Cayman Chemical Supplier No. 4028010 MG
A lignan with diverse biological activities; scavenges DPPH and ABTS radicals in cell-free assays (IC50s = 9.9 and 12.7 µM, respectively); inhibits LPS-induced NO production in RAW 264.7 cells at 20 µM; reverses scopolamine-induced memory impairment, reduces hippocampal MDA levels, and increases hippocampal SOD, catalase, and GPX levels in a mouse model of scopolamine-induced amnesia at 20 mg/kg