Learn More
Biomatik Corporation Aco1 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11187
Encompass_Preferred
Aco1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ACO1. The Aco1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Cytoplasmic Aconitate Hydratase Protein (508-745Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: ACO1. Accession Number: P21399. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9248