Learn More
Sino Biological ACO2 / Aconitase 2 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 10958T60100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ACO2 / Aconitase 2 (rh ACO2 / Aconitase 2; Catalog#10958-H20B; Q99798; Gln28-Gln780). ACO2 / Aconitase 2 specific IgG was purified by Human ACO2 / Aconitase 2 affinity chromatography.
Catalog No. 50-224-7941