Learn More
Synbiosis™ aCOLyte Instrument Validation Procedures
GREENER_CHOICESDP
Catalog No. 01257339 Shop All Synoptics, Inc. Products
Change view
Click to view available options
Service Offering:
Installation, Operational, and Performance Qualification
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Service Offering
01-257-339 -
01-257-340 Installation, Operational, and Performance Qualification
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-257-339 Supplier Synbiosis™ Supplier No. IQOQAC
Add to Cart
Add to Cart
aCOLyte Instrument Validation Procedures take the user through each step in the verification of a Synbiosis™ system regulatory compliances and standard operational procedures (SOP). These simple procedures allow the user to verify that a system meets defined installation, operational, and performance quality specifications.
  • Installation Qualification (IQ): Verifies that when a system is unpacked it conforms with all purchasing, shipping, and unpacking documents. Once this procedure has been followed the system is then ready for the operational check process.
  • Operational Qualification (OQ): Establishes if the system meets all the operation and functionality specifications. An operational test is also included at this stage.
  • Performance Qualification (PQ): Establishes if the system meets the specification for the nature of the purpose it was intended for. This is verified by using a simple target to establish if the system can meet performance criteria.
  • Documentation: Each set of IQ/OQ/PQ documents contains a complete set of procedures and checklists for following the verification steps. Included with each pack is a simple calibration device (target image) so that the performance of the system can be checked against the PQ requirements. Each set of documents is unique to a particular system and has to be ordered with the system itself. The document pack has full instructions for use and implementation.
Specifications
For Use With (Equipment) aCOLyte Instruments
For Use With (Application) Instrument Validation
WARNING: Cancer - www.P65Warnings.ca.gov