Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synbiosis™ aCOLyte Sample Holder for Oxoid Plates, 80 mm
GREENER_CHOICESDP

Catalog No. 01257336 Shop All Synoptics, Inc. Products
Change view
Click to view available options
For Use With (Application):
Holding oxoid plates, 80 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
01-257-336 Holding oxoid plates, 80 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-257-336 Supplier Synbiosis™ Supplier No. ACOLPHOX
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

aCOLyte Sample Holder is designed for use with 80 mm oxoid plates.

  • Holds 80 mm oxoid plates
  • For use with aCOLyte 3 instruments

Specifications

Product Type Plate Holder
Material Plastic
Product Line Synbiosis
For Use With (Application) Holding oxoid plates, 80 mm
For Use With (Equipment) aCOLyte Instruments
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.