Learn More
Medchemexpress LLC Aconitane-8,13,14,15-tetrol, 1,6,16-trimethoxy-4-(methoxymethyl)-20-methyl | 63238-68-6 | 469.57 | 1 MG
SDP
Catalog No. 5000415051
Encompass_Preferred
Hypaconine is a C19-diterpenoid alkaloid isolated from *Aconitum* and *Delphinium* species. It exhibits strong cardiac activity and is intended for research use only, not for medical applications or sale to patients.
  • C19-diterpenoid alkaloid
  • Isolated from *Aconitum* and *Delphinium* species
  • Exhibits strong cardiac activity
  • For research use only
Catalog No. 50-004-15051 Supplier Medchemexpress LLC Supplier No. HYN19231MG
Hypaconine is a C19-diterpenoid alkaloid isolated from *Aconitum* and *Delphinium* species. It exhibits strong cardiac activity and is intended for research use only, not for medical applications or sale to patients.
  • C19-diterpenoid alkaloid
  • Isolated from *Aconitum* and *Delphinium* species
  • Exhibits strong cardiac activity
  • For research use only