Learn More
Cayman Chemical Aconitase Assay Kit, 96 Well
SDP
Catalog No. 502894227
Encompass_Preferred
Aconitase is an iron-sulfur protein containing a [Fe4S4]2+ cluster that catalyzes the stereospecific isomerization of citrate to isocitrate via cis-aconitate. Whereas exposure of aconitase to oxidants renders the enzyme inactive, loss of aconitase activity in cells or in biological samples treated with pro-oxidants has been interpreted as a measure of oxidative damage. Cayman’s Aconitase Assay provides a simple, reproducible, and sensitive tool for assaying aconitase from tissue homogenates or cell lysates.
Catalog No. 50-289-4227 Supplier Cayman Chemical Supplier No. 70550296 WELL
Aconitase is an iron-sulfur protein containing a [Fe4S4]2+ cluster that catalyzes the stereospecific isomerization of citrate to isocitrate via cis-aconitate. Whereas exposure of aconitase to oxidants renders the enzyme inactive, loss of aconitase activity in cells or in biological samples treated with pro-oxidants has been interpreted as a measure of oxidative damage. Cayman’s Aconitase Assay provides a simple, reproducible, and sensitive tool for assaying aconitase from tissue homogenates or cell lysates.