Learn More
Cayman Chemical AconItase IsocItrIc Dehydroge
SDP
Catalog No. 502893263
Encompass_Preferred
A Bax activator (Ki 43.3 nM in a fluorescence polarization assay) selectively reduces viability in wild-type but not Bax-/- MEFs at 25 UM inhibits nicotine-induced increases in phosphorylated Bax levels in A549 cells at 5 UM reduces viability in A549 cells at 5 UM induces cell cycle arrest at the G2/M phase in U87MG cells at 5 10 or 15 UM disrupts the mitochondrial membrane potential in U87MG cells at 15 UM induces cytochrome c release from mitochondria and Bax insertion into mitochondrial membranes in A549 cells at 5 or 10 UM decreases tumor volume in an A549 mouse xenograft model at 40 or 60 mg/kg per day
Catalog No. 50-289-3263 Supplier Cayman Chemical Supplier No. 7055101 EA
A Bax activator (Ki 43.3 nM in a fluorescence polarization assay) selectively reduces viability in wild-type but not Bax-/- MEFs at 25 UM inhibits nicotine-induced increases in phosphorylated Bax levels in A549 cells at 5 UM reduces viability in A549 cells at 5 UM induces cell cycle arrest at the G2/M phase in U87MG cells at 5 10 or 15 UM disrupts the mitochondrial membrane potential in U87MG cells at 15 UM induces cytochrome c release from mitochondria and Bax insertion into mitochondrial membranes in A549 cells at 5 or 10 UM decreases tumor volume in an A549 mouse xenograft model at 40 or 60 mg/kg per day