Learn More
Enzo Life Sciences Aconitine (25mg). CAS: 302-27-2
SDP
Supplier:  Enzo Life Sciences ALX550232M025
Encompass_Preferred
A lipid-soluble neurotoxin that acts as a sodium channel modulator. Binds with high affinity to the open state of the voltage-sensitive sodium channels at site 2, thereby causing a persistent activation of the sodium channels, which become refractory to excitation. Alternative name: 1alpha,3alpha,6alpha,14alpha,15alpha,16beta)-20-Ethyl-1,6,16-trimethoxy-4-(methoxymethyl)aconitane-3,8,13,14,15-pentol 8-acetate 14-benzoate. Purity: ≥98% (HPLC). Solubility: Soluble in 100% ethanol (35mg/ml); almost insoluble in water (0.3mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-9503