Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aconitine, 98%

Catalog No. AAJ62056MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62056MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62056MD Supplier Thermo Scientific Chemicals Supplier No. J62056MD
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 302-27-2 | C34H47NO11 | 645.746 g/mol

Opens tetrodotoxin-sensitive sodium channels. Useful for creating models of cardiac arrhythmia

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Opens tetrodotoxin-sensitive sodium channels
  • Useful for creating models of cardiac arrhythmia
TRUSTED_SUSTAINABILITY

Specifications

CAS 302-27-2
Melting Point 194°C to 202°C
Color White
Molecular Formula C34H47NO11
Quantity 250 mg
MDL Number MFCD00082375
UN Number UN1544
Beilstein 74608
Merck Index 14,118
Synonym aconitine
Solubility Information Soluble in ethanol; Sparingly soluble in water
InChI Key XFSBVAOIAHNAPC-OZUFZMTGSA-N
SMILES CCN1CC2(C(CC(C34C2C(C(C31)C5(C6C4CC(C6OC(=O)C7=CC=CC=C7)(C(C5O)OC)O)OC(=O)C)OC)OC)O)COC
Molecular Weight (g/mol) 645.746
PubChem CID 132470875
Formula Weight 645.75
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Aconitine
Show More Show Less
Hazard Category H300+H330
Hazard Statement GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.
Precautionary Statement P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S.
EINECSNumber 206-121-7
RTECSNumber AR5960000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.