Learn More
Cayman Chemical aContoxIn PnIAtrIfluoroac 10mg
SDP
Catalog No. 502911946
Encompass_Preferred
A peptide antagonistic of α7 and α3β2-subunit containing nAChRs (IC50s = 62.7 and 7.9 nM, respectively); inhibits tumor growth and increases survival in an Ehrlich murine spontaneous adenocarcinoma model at 0.1 nmole/kg in combination with baicalein
Catalog No. 50-291-1946 Supplier Cayman Chemical Supplier No. 3619710 MG
A peptide antagonistic of α7 and α3β2-subunit containing nAChRs (IC50s = 62.7 and 7.9 nM, respectively); inhibits tumor growth and increases survival in an Ehrlich murine spontaneous adenocarcinoma model at 0.1 nmole/kg in combination with baicalein