Learn More
Cayman Chemical Acontum carmchal Dlsoln 10mg
SDP
Catalog No. 502991499
Encompass_Preferred
A diterpene alkaloid with diverse biological activities; inhibits binding of α-bungarotoxin to rat neuronal α7 nAChRs (IC50 = 19 µM); repellent against adult red flour beetles in a filter paper assay at 3 mg/ml; lowers systolic blood pressure in a rat model of left renal artery ligation-induced hypertension at 75 mg/kg
Catalog No. 50-299-1499 Supplier Cayman Chemical Supplier No. 3174810 MG
A diterpene alkaloid with diverse biological activities; inhibits binding of α-bungarotoxin to rat neuronal α7 nAChRs (IC50 = 19 µM); repellent against adult red flour beetles in a filter paper assay at 3 mg/ml; lowers systolic blood pressure in a rat model of left renal artery ligation-induced hypertension at 75 mg/kg