Learn More
Cayman Chemical Acontum carmchal Dltaln 10mg
SDP
Catalog No. 502949674
Encompass_Preferred
An alkaloid and a toxin; reduces sciatic nerve stimulation-induced CMAPS in isolated lizard extensor digitorum longus muscle (IC50 = 156 µM); induces neuromuscular impairment in mice at 85 mg/kg; induces lethality in mice (LD50 = 113 mg/kg)
Catalog No. 50-294-9674 Supplier Cayman Chemical Supplier No. 3461210 MG
An alkaloid and a toxin; reduces sciatic nerve stimulation-induced CMAPS in isolated lizard extensor digitorum longus muscle (IC50 = 156 µM); induces neuromuscular impairment in mice at 85 mg/kg; induces lethality in mice (LD50 = 113 mg/kg)