Learn More
Cayman Chemical Denudatine, Purity: ≥95%, CAS: 26166-37-0, 10 mg
SDP
Catalog No. 502991375
Encompass_Preferred
An alkaloid with antiarrhythmic activities; prevents aconitine-induced arrhythmias in rats and increases survival in a rat model of calcium chloride-induced arrhythmias at 50 and 100 mg/kg
Catalog No. 50-299-1375 Supplier Cayman Chemical Supplier No. 3090410 MG
An alkaloid with antiarrhythmic activities; prevents aconitine-induced arrhythmias in rats and increases survival in a rat model of calcium chloride-induced arrhythmias at 50 and 100 mg/kg