Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acoramidis hydrochloride | 2242751-53-5 | 98.5% | 328.77 | 25 MG
SDP

Catalog No. 5000347391
Encompass_Preferred

Acoramidis hydrochloride is an orally active and selective kinetic stabilizer of WT and V122I-TTR (transthyretin). It is used in the study for transthyretin amyloidosis.

  • Solid appearance
  • Color white to light yellow
  • Soluble in DMSO (62.5 mg/mL), insoluble in H2O
  • Stabilizes V122I- and WT-TTR
  • Stimulates mitochondrial QO2
  • Shows minimal inhibition of hERG and cytochrome P450 isozymes

Catalog No. 50-003-47391 Supplier Medchemexpress LLC Supplier No. 25MGHY109165A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Acoramidis hydrochloride is an orally active and selective kinetic stabilizer of WT and V122I-TTR (transthyretin). It is used in the study for transthyretin amyloidosis.

  • Solid appearance
  • Color white to light yellow
  • Soluble in DMSO (62.5 mg/mL), insoluble in H2O
  • Stabilizes V122I- and WT-TTR
  • Stimulates mitochondrial QO2
  • Shows minimal inhibition of hERG and cytochrome P450 isozymes

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.