Learn More
Medchemexpress LLC Acoramidis hydrochloride | 2242751-53-5 | 98.7% | 328.77 | 50 MG
SDP
Catalog No. 5000344142
Encompass_Preferred
Acoramidis (AG10) hydrochloride is an orally active and selective kinetic stabilizer of WT and V122I-TTR (transthyretin). It is utilized in the study of transthyretin amyloidosis. This compound demonstrates equal efficacy in stabilizing V122I- and WT-TTR and surpasses tafamidis in stabilizing both WT and mutant TTR in whole serum. Additionally, it exhibits minimal inhibition of common drug discovery off-targets like the hERG potassium ion channel and cytochrome P450 isozymes, indicating low toxicity.
  • Stabilizes WT and V122I-TTR.
  • Used in the study for transthyretin amyloidosis.
  • Shows minimal inhibition of hERG and cytochrome P450 isozymes, suggesting low toxicity.
Catalog No. 50-003-44142 Supplier Medchemexpress LLC Supplier No. HY109165A50MG
Acoramidis (AG10) hydrochloride is an orally active and selective kinetic stabilizer of WT and V122I-TTR (transthyretin). It is utilized in the study of transthyretin amyloidosis. This compound demonstrates equal efficacy in stabilizing V122I- and WT-TTR and surpasses tafamidis in stabilizing both WT and mutant TTR in whole serum. Additionally, it exhibits minimal inhibition of common drug discovery off-targets like the hERG potassium ion channel and cytochrome P450 isozymes, indicating low toxicity.
  • Stabilizes WT and V122I-TTR.
  • Used in the study for transthyretin amyloidosis.
  • Shows minimal inhibition of hERG and cytochrome P450 isozymes, suggesting low toxicity.