Learn More
TARGETMOL CHEMICALS INC ACORAMIDIS HYDROCHLORIDE 5MG
SDP
Catalog No. 502580036
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Acoramidis hydrochloride (Alxn2060 hydrochloride) is an orally active and selective kinetic stabilizer of WT and V122I- TTR (transthyretin) . Acoramidis hydrochloride is used in the study for transthyretin amyloidosis. purity: 99%
Catalog No. 50-258-0036 Supplier TARGETMOL CHEMICALS INC Supplier No. T397405MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Acoramidis hydrochloride (Alxn2060 hydrochloride) is an orally active and selective kinetic stabilizer of WT and V122I- TTR (transthyretin) . Acoramidis hydrochloride is used in the study for transthyretin amyloidosis. purity: 99%