Learn More
Cayman Chemical Acoramidis (hydrochloride), Purity: ≥98%, CAS: 2242751-53-5, 10 mg
SDP
Catalog No. 502944414
Encompass_Preferred
A TTR kinetic stabilizer; selectively binds to wild-type TTR and TTRV122I with negative cooperativity (Kd1s = 4.8 and 6.2 nM, respectively; Kd2s = 314 and 139 nM, respectively) over COX-1, COX-2, and the thyroid hormone nuclear receptor (IC50s = >10 µM for all); inhibits pH-induced amyloidogenesis of wild-type TTR and TTRV122I, indicating tetramer stabilization; stabilizes TTRV122I in serum isolated from patients with familial amyloid cardiomyopathy at 10 µM
Catalog No. 50-294-4414 Supplier Cayman Chemical Supplier No. 4153610 MG
A TTR kinetic stabilizer; selectively binds to wild-type TTR and TTRV122I with negative cooperativity (Kd1s = 4.8 and 6.2 nM, respectively; Kd2s = 314 and 139 nM, respectively) over COX-1, COX-2, and the thyroid hormone nuclear receptor (IC50s = >10 µM for all); inhibits pH-induced amyloidogenesis of wild-type TTR and TTRV122I, indicating tetramer stabilization; stabilizes TTRV122I in serum isolated from patients with familial amyloid cardiomyopathy at 10 µM