Learn More
Aviva Systems Biology Corp ACOT11 Antibody - middle region (ARP55281_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55281_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACOT11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ACOT11. Alternate names: BFIT, BFIT1, BFIT2, KIAA0707, STARD14, THEA, THEM1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61455