Learn More
Aviva Systems Biology Corp ACOT2 Antibody - middle region (ARP42322_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42322_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACOT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ACOT2. Alternate names: Mte1, PTE2, ZAP128, MTE1, CTE1A, PTE2AApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58182