Learn More
Aviva Systems Biology Corp ACOT8 Antibody - N-terminal region (ARP62050_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62050_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACOT8. It was validated on Western Blot. . Alternate names: HNAACTE, PTE-2, PTE1, PTE2, hACTE-III, hTE, PTE-1ApplicationWB. Species reactivity: Human
Catalog No. 50-287-552