Learn More
Aviva Systems Biology Corp ACOT9 Antibody - C-terminal region (ARP60816_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60816_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACOT9. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ACOT9. Alternate names: ACATE2, CGI-16, MT-ACT48, MTACT48ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Rat
Catalog No. 50-287-177