Learn More
Selleck Chemical LLC Acotiamide hydrochloride S4838-1g
SDP
Catalog No. 503108986
Encompass_Preferred
Acotiamide Hydrochloride (YM-443 Z-338) is the hydrochloride salt form of acotiamide a prokinetic agent with gastrointestinal (GI) motility-enhancing activity It is a new orally active selective acetylcholinesterase inhibitor
Catalog No. 50-310-8986 Supplier Selleck Chemical LLC Supplier No. S48381G
Acotiamide Hydrochloride (YM-443 Z-338) is the hydrochloride salt form of acotiamide a prokinetic agent with gastrointestinal (GI) motility-enhancing activity It is a new orally active selective acetylcholinesterase inhibitor