Learn More
Medchemexpress LLC Acotiamide hydrochloride | 185104-11-4 | 99.6% | 10 MG
SDP
Catalog No. 5000231939
Encompass_Preferred
Acotiamide hydrochloride is a research-grade small molecule and reversible acetylcholinesterase (AChE) inhibitor (IC50 1.79 μM) used to study gastric motility, delayed gastric emptying, and functional dyspepsia. Supplied as the hydrochloride salt with high reported purity for biochemical and pharmacological research.
  • Selective, reversible AChE inhibitor with reported IC50 1.79 μM.
  • Suitable for studies of gastric motility and functional dyspepsia models.
  • Reported high purity (99.62%), appropriate for assay use.
  • Storage: sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Provided in small milligram pack sizes for laboratory research.
Catalog No. 50-002-31939 Supplier Medchemexpress LLC Supplier No. HY121467A10MG
Acotiamide hydrochloride is a research-grade small molecule and reversible acetylcholinesterase (AChE) inhibitor (IC50 1.79 μM) used to study gastric motility, delayed gastric emptying, and functional dyspepsia. Supplied as the hydrochloride salt with high reported purity for biochemical and pharmacological research.
  • Selective, reversible AChE inhibitor with reported IC50 1.79 μM.
  • Suitable for studies of gastric motility and functional dyspepsia models.
  • Reported high purity (99.62%), appropriate for assay use.
  • Storage: sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Provided in small milligram pack sizes for laboratory research.