Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acotiamide hydrochloride | 185104-11-4 | 99.6% | 25 MG
SDP

Catalog No. 5000233681
Encompass_Preferred

Acotiamide hydrochloride is the hydrochloride salt of acotiamide, provided as a research-grade solid reagent used to study gastric motility, delayed gastric emptying, and related gastrointestinal disorders. It is supplied in small pack sizes with high reported purity suitable for analytical and in vitro research.

  • Orally active, reversible acetylcholinesterase inhibitor (reported IC50 ~1.79 μM).
  • Hydrochloride salt for improved stability and solubility.
  • High reported purity for research applications (reported ~99.6%).
  • Molecular formula C21H31ClN4O5S; molecular weight 487.01.
  • Available in small research pack sizes such as 25 mg.

Catalog No. 50-002-33681 Supplier Medchemexpress LLC Supplier No. HY121467A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acotiamide hydrochloride is the hydrochloride salt of acotiamide, provided as a research-grade solid reagent used to study gastric motility, delayed gastric emptying, and related gastrointestinal disorders. It is supplied in small pack sizes with high reported purity suitable for analytical and in vitro research.

  • Orally active, reversible acetylcholinesterase inhibitor (reported IC50 ~1.79 μM).
  • Hydrochloride salt for improved stability and solubility.
  • High reported purity for research applications (reported ~99.6%).
  • Molecular formula C21H31ClN4O5S; molecular weight 487.01.
  • Available in small research pack sizes such as 25 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.