Learn More
Medchemexpress LLC Acotiamide monohydrochloride trihydrate | 773092-05-0 | 99.8% | 1 G
SDP
Catalog No. 5000411602
Encompass_Preferred
Acotiamide monohydrochloride trihydrate is an orally active, selective, and reversible acetylcholinesterase (AChE) inhibitor with an IC50 of 1.79 μM. It can enhance gastric contractility and accelerate delayed gastric emptying. It has potential for research into functional dyspepsia involving gastric motility dysfunction and intestinal inflammatory conditions.
  • Orally active, selective, and reversible acetylcholinesterase (AChE) inhibitor
  • Enhances gastric contractility and accelerates delayed gastric emptying
  • Potential for research of functional dyspepsia and intestinal inflammatory conditions
Catalog No. 50-004-11602 Supplier Medchemexpress LLC Supplier No. HYB21551G
Acotiamide monohydrochloride trihydrate is an orally active, selective, and reversible acetylcholinesterase (AChE) inhibitor with an IC50 of 1.79 μM. It can enhance gastric contractility and accelerate delayed gastric emptying. It has potential for research into functional dyspepsia involving gastric motility dysfunction and intestinal inflammatory conditions.
  • Orally active, selective, and reversible acetylcholinesterase (AChE) inhibitor
  • Enhances gastric contractility and accelerates delayed gastric emptying
  • Potential for research of functional dyspepsia and intestinal inflammatory conditions