Learn More
Medchemexpress LLC Acotiamide monohydrochloride trihydrate | 773092-05-0 | 99.8% | 500 MG
SDP
Catalog No. 5000411500
Encompass_Preferred
Acotiamide monohydrochloride trihydrate is an orally active, selective, and reversible acetylcholinesterase inhibitor, with an IC50 of 1.79 μM. It can enhance gastric contractility and accelerate delayed gastric emptying. This compound has potential for the research of functional dyspepsia involving gastric motility dysfunction and intestinal inflammatory conditions.
  • Acts as an orally active, selective, and reversible acetylcholinesterase inhibitor.
  • Enhances gastric contractility.
  • Accelerates delayed gastric emptying.
  • Has potential for research into functional dyspepsia involving gastric motility dysfunction and intestinal inflammation.
Catalog No. 50-004-11500 Supplier Medchemexpress LLC Supplier No. HYB2155500MG
Acotiamide monohydrochloride trihydrate is an orally active, selective, and reversible acetylcholinesterase inhibitor, with an IC50 of 1.79 μM. It can enhance gastric contractility and accelerate delayed gastric emptying. This compound has potential for the research of functional dyspepsia involving gastric motility dysfunction and intestinal inflammatory conditions.
  • Acts as an orally active, selective, and reversible acetylcholinesterase inhibitor.
  • Enhances gastric contractility.
  • Accelerates delayed gastric emptying.
  • Has potential for research into functional dyspepsia involving gastric motility dysfunction and intestinal inflammation.