Learn More
Grainger Auralex Acoustics Acoustic Foam, Polyurethane, 4 in. Thick, 6/Pk.
Catalog No. 502779065
Encompass_Preferred
These acoustic foam panels are designed to effectively reduce sound reflections and improve sonic accuracy in professional and home environments. Constructed from high-density, open-celled polyurethane foam, each 4 in. thick panel features a wedge pattern that maximizes acoustic absorption, particularly in rooms with pronounced low-frequency challenges. These versatile panels are suitable for applications in concert halls, rehearsal spaces, and churches, and they maintain their physical integrity over time without crumbling or fading.
  • Provides a noise reduction coefficient (NRC) of 1.1
  • Includes six 4 in. x 24 in. x 24 in. panels
  • Engineered for flexible wall or ceiling mounting
  • Constructed from flame-retardant polyurethane foam
  • Meets flammability standards including UL-94 HF-1 and CAL 117
  • Compliant with REACH and RoHS regulations
Catalog No. 50-277-9065 Supplier Grainger Supplier No. 19MP20
These acoustic foam panels are designed to effectively reduce sound reflections and improve sonic accuracy in professional and home environments. Constructed from high-density, open-celled polyurethane foam, each 4 in. thick panel features a wedge pattern that maximizes acoustic absorption, particularly in rooms with pronounced low-frequency challenges. These versatile panels are suitable for applications in concert halls, rehearsal spaces, and churches, and they maintain their physical integrity over time without crumbling or fading.
  • Provides a noise reduction coefficient (NRC) of 1.1
  • Includes six 4 in. x 24 in. x 24 in. panels
  • Engineered for flexible wall or ceiling mounting
  • Constructed from flame-retardant polyurethane foam
  • Meets flammability standards including UL-94 HF-1 and CAL 117
  • Compliant with REACH and RoHS regulations