Learn More
Grainger Auralex Studiofoam SonoMatt Acoustic Foam Panel, Eggcrate, Charcoal, 2/Pk.
Catalog No. 502782493
Encompass_Preferred
These acoustic foam panels utilize an eggcrate pattern for effective broadband sound absorption in rooms when applied to walls or ceilings. Constructed from open cell polyurethane foam, they provide a cost-effective solution for acoustical management while maintaining reliable performance.
  • Features an industry-standard eggcrate pattern for optimal sound absorption
  • Constructed from durable open cell polyurethane foam
  • Certified for flammability under CAL 117 and ASTM-D3574 standards
  • Available in a charcoal finish
  • Compatible with standard mounting methods including foam-specific adhesives and tabs
  • Provides an overall noise reduction coefficient (NRC) of 0.70
Catalog No. 50-278-2493 Supplier Grainger Supplier No. 19MP18
These acoustic foam panels utilize an eggcrate pattern for effective broadband sound absorption in rooms when applied to walls or ceilings. Constructed from open cell polyurethane foam, they provide a cost-effective solution for acoustical management while maintaining reliable performance.
  • Features an industry-standard eggcrate pattern for optimal sound absorption
  • Constructed from durable open cell polyurethane foam
  • Certified for flammability under CAL 117 and ASTM-D3574 standards
  • Available in a charcoal finish
  • Compatible with standard mounting methods including foam-specific adhesives and tabs
  • Provides an overall noise reduction coefficient (NRC) of 0.70