Learn More
Grainger Auralex ProMAX V2 Acoustic Panel, Charcoal, Portable, 2/Pk.
Catalog No. 502779760
Encompass_Preferred
These portable acoustic absorption devices provide a flexible solution for managing sound reflections and room modes in professional and residential environments. Designed for situations where wall-mounted treatments are not feasible, these units can be easily positioned on included stands to improve acoustic performance in studios, classrooms, and stage settings.
  • Lightweight and portable design for versatile placement
  • Highly effective at controlling axial modes and taming chaotic reverberation
  • Includes two 2 ft x 4 ft panels with 3 in depth
  • Features a semi-reflective, angled rear surface to manage reflections
  • Adjustable height provided by included round base stands
  • Certified fire resistant per ASTM D3574 and CAL 117 standards
  • Noise reduction coefficient of 0.95
Catalog No. 50-277-9760 Supplier Grainger Supplier No. 19MP54
These portable acoustic absorption devices provide a flexible solution for managing sound reflections and room modes in professional and residential environments. Designed for situations where wall-mounted treatments are not feasible, these units can be easily positioned on included stands to improve acoustic performance in studios, classrooms, and stage settings.
  • Lightweight and portable design for versatile placement
  • Highly effective at controlling axial modes and taming chaotic reverberation
  • Includes two 2 ft x 4 ft panels with 3 in depth
  • Features a semi-reflective, angled rear surface to manage reflections
  • Adjustable height provided by included round base stands
  • Certified fire resistant per ASTM D3574 and CAL 117 standards
  • Noise reduction coefficient of 0.95