Learn More
Grainger Auralex Acoustical Sealant, Latex Based, 28 oz., 1 ea.
Catalog No. 502779268
Encompass_Preferred
StopGap™ Acoustical Sealant is a highly elastic, water-based paste designed to minimize sound transmission at the perimeter of wall systems and around electrical box cutouts. This professional-grade sealant helps achieve specified STC and MTC values in soundproofing applications and cures to a durable, opaque white film.
  • Remains flexible and stays pliable for up to five times longer than alternative brands
  • Provides a non-staining, fully paintable finish
  • Meets ASTM E-84 and ASTM C-834 standards for flame retardancy
  • Ensures clean application with no residual run-off
  • Requires a larger than normal-sized caulking gun due to 29 oz. tube size
Catalog No. 50-277-9268 Supplier Grainger Supplier No. 19MP46
StopGap™ Acoustical Sealant is a highly elastic, water-based paste designed to minimize sound transmission at the perimeter of wall systems and around electrical box cutouts. This professional-grade sealant helps achieve specified STC and MTC values in soundproofing applications and cures to a durable, opaque white film.
  • Remains flexible and stays pliable for up to five times longer than alternative brands
  • Provides a non-staining, fully paintable finish
  • Meets ASTM E-84 and ASTM C-834 standards for flame retardancy
  • Ensures clean application with no residual run-off
  • Requires a larger than normal-sized caulking gun due to 29 oz. tube size