Learn More
Aviva Systems Biology Corp ACOX3 Antibody - C-terminal region (ARP60790_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60790_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACOX3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ACOX3. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-287-164