Learn More
Medchemexpress LLC HY-19910 5mg Medchemexpress, Acoziborole CAS:1266084-51-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY199105MG
Encompass_Preferred
Medchemexpress, HY-19910 5mg Acoziborole CAS:1266084-51-8 Acoziborole (SCYX-7158) is an effective, safe and orally active antiprotozoal agent for the research of human african trypanosomiasis (HAT). In the T. b. brucei S427 strain, the MIC value for SCYX-7158 is 0.6 µg/mL. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2197